Tlačiť

Anti-LTBR Rabbit Monoclonal Antibody [clone: EPR26096-171]

Dodávateľ: ABCAM
AB313561-100UL AB313561-20UL AB313561-1ML
ABCAAB313561-100ULEA 793.42 EUR
ABCAAB313561-100UL ABCAAB313561-20UL ABCAAB313561-1ML
Anti-LTBR Rabbit Monoclonal Antibody [clone: EPR26096-171]
Protilátky
Rabbit monoclonal [EPR26096-171] to LTBR.

Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: LTBR
Clonality: Monoclonal
Clone: EPR26096-171
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR