Tlačiť
Anti-IRE1 (phospho S724) Rabbit Monoclonal Antibody [clone: EPR23107-16]
Dodávateľ: ABCAM
AB267395-100 AB267395-1
ABCAAB267395-100EA 1164.86 EUR
ABCAAB267395-100 ABCAAB267395-1
Anti-IRE1 (phospho S724) Rabbit Monoclonal Antibody [clone: EPR23107-16]
Protilátky
Rabbit monoclonal [EPR23107-16] to IRE1 (phospho S724) - BSA and Azide free.
Pozor: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: IRE1
Clonality: Monoclonal
Clone: EPR23107-16
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human