Tlačiť
Capillary columns, PERMABOND® SE-30
Dodávateľ: MACHEREY-NAGEL
723052.25 723052.50 723306.25 723308.50
MANA723052.25EA 761.73 EUR
MANA723052.25 MANA723052.50 MANA723306.25 MANA723308.50
Capillary columns, PERMABOND® SE-30
Kolony chromatografické
  • Nonpolar phase
  • Maximum temperature 300 °C (long-term temperature)
  • Maximum temperature 320 °C (short-term max. temperature in a temperature program)