Tlačiť
Capillary columns, OPTIMA® 240
Dodávateľ: MACHEREY-NAGEL
726089.30 726089.50 726089.60 726090.30 726090.60 726091.25 726091.30 726091.50 726091.60 726095.30 726095.60 726096.30 726096.60
MANA726089.30EA 761.73 EUR
MANA726089.30 MANA726089.50 MANA726089.60 MANA726090.30 MANA726090.60 MANA726091.25 MANA726091.30 MANA726091.50 MANA726091.60 MANA726095.30 MANA726095.60 MANA726096.30 MANA726096.60
Capillary columns, OPTIMA® 240
Kolony chromatografické
Medium polar GC phase.
  • Recommended for FAMEs, and dioxins
  • Maximum temperature for isothermal operation 260 °C
  • Maximum temperature for short isotherms in a temperature program 280 °C
33% cyanopropyl-methyl – 67% dimethylpolysiloxane phase.