Tlačiť
Capillary columns, OPTIMA® 624
Dodávateľ: MACHEREY-NAGEL
726784.25 726785.25 726785.30 726785.50 726785.60 726787.25 726787.30 726787.50 726787.60 726789.25 726789.30 726786.30 726786.50
MANA726784.25EA 761.73 EUR
MANA726784.25 MANA726785.25 MANA726785.30 MANA726785.50 MANA726785.60 MANA726787.25 MANA726787.30 MANA726787.50 MANA726787.60 MANA726789.25 MANA726789.30 MANA726786.30 MANA726786.50
Capillary columns, OPTIMA® 624
Kolony chromatografické
Medium polar GC phases.
  • Recommended for environmental analyses such as halogenated hydrocarbons, and volatiles as well as for solvents
  • Maximum temperature for isothermal operation 280 °C
  • Maximum temperature for short isotherms in a temperature program 300 °C
6% cyanopropyl-phenyl – 94% dimethylpolysiloxane phase.
USP code: G43.