Tlačiť
Basic Laboratory Chairs
Dodávateľ: BIMOS SITZTECHNIK
9131-6801 9138-6902 9131-6911 9133-2571 9134-2571 9134-6902 9134-6911 9138-2571
BIMO9131-6801EA 548.34 EUR
BIMO9131-6801 BIMO9138-6902 BIMO9131-6911 BIMO9133-2571 BIMO9134-2571 BIMO9134-6902 BIMO9134-6911 BIMO9138-2571
Basic Laboratory Chairs
Židle
The tried-and-tested all-rounder for use in laboratories is a great entry level laboratory chair, perfect for a range of workplace environments.
  • With a sturdy steel 5 star base
  • Available in two different frame colours: black or light grey
  • Choice of three different mechanisms: contact backrest, contact backrest with seat tilt adjustment and synchronous mechanism with weight regulation
Basic Laboratory can be supplied with Stamskin synthetic leather or with fabric upholstery. Not only is Stamskin Top easy to clean, but it also has antibacterial, antifungal and antimicrobial properties.
Conform to: DIN 68 877, German GS „safety tested“ certificate and Quality Management System DIN EN ISO 9001
Certifikácie: Conform to: DIN 68 877, German GS 'safety tested' certificate and Quality Management System DIN EN ISO 9001