Tlačiť
Sleeve protectors, cut resistant
Dodávateľ: Ansell
70-114 70-118 70-123 70-110
113-8690EA 10.11 EUR
113-8690 112-2932 112-2934 112-2930
Sleeve protectors, cut resistant
Pracovní oděvy
These knitted liners are constructed from 100% Kevlar® para-aramid short fibres.
  • Exceptional cut resistance with dexterity
  • Most models feature thumb slots, protecting the wrist area
They are available in different versions and lengths. All sleeves provide superior mechanical protection and cut resistance.
Gauge: 24
Certifikácie: EN 388 (134X), EN 407 (X1XXXX); Cat. II