Tlačiť

Etidronic acid monohydrate, Sigma-Aldrich®

Dodávateľ: Merck

54342-10G 54342-50G
SIAL54342-10GEA 156.04 EUR
SIAL54342-10G SIAL54342-50G
Etidronic acid monohydrate, Sigma-Aldrich®
Etidronic acid monohydrate
Vzorec: C₂H₈O₇P₂·1H₂O
Číslo CAS: 25211-86-3
EINECS: 220-552-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR