Tlačiť
Solid phase extraction cartridges, Bond Elut™ AccuCAT
Dodávateľ: VARIAN
12282001 12282003 12282004 12282002 12282005 12282006
VARI12282001EA 353.39 EUR
VARI12282001 551-0417 VARI12282004 VARI12282002 VARI12282005 VARI12282006
Solid phase extraction cartridges, Bond Elut™ AccuCAT
Kolony chromatografické
Bond Elut AccuCAT cartridges are mixed bed SPE cartridges consisting of a strong cation exchange (SCX) and a strong anion exchange (SAX) sorbent packed into one bed. AccuCAT is effective for the extraction of acidic, basic and neutral analytes from urine and other biological samples. AccuCAT is particularly effective for catecholamine extraction from bio fluids.
  • Ultra clean, mixed sorbent bed delivers for reproducible extractions
  • SCX and SAX functionalities offer broad analyte extraction potential
  • Compatible with many biological fluids for easy method transfer