Tlačiť
PIG® spill kit, 115 L overpack
Dodávateľ: NEW PIG
KIT236 KIT336 KIT436 RFL236 RFL336 RFL436
121-0554EA 367.83 EUR
121-0554 121-0055 121-0058 121-0555 121-0113 121-0115
PIG® spill kit, 115 L overpack
Absorpční materiály
Strong enough to earn a UN rating for transporting spill cleanup waste.
  • Comply with spill plan regs, avoid fines and be ready to respond
  • Overpack drum container is UN-rated (UN1H2/X70/S) for transporting waste after spill cleanup
  • Neatly prepacked container speeds access during a spill emergency
  • Overpack is X-rated in Packing Groups I, II and III for transporting spill cleanup waste by land, sea or rail
  • Lightweight polypropylene container resists chemicals and keeps contents clean and dry
  • Two broom holes 180-degrees opposite each other on the lid allow 100% positive leverage for easy opening and closing with a broom handle or bar
The ledges molded into the container make kit easy to move by forklift; foot hold in the drum base allows you to hold the drum securely in position while opening or closing the lid.
Pozor: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.