Tlačiť

N-Acetonylpyridinium chloride ≥97%

Dodávateľ: Thermo Fisher Scientific
Varování

Synonymá: 1-Acetonylpyridiniumchloride, 1-Acetonylpyridinium chloride

A16015.06 A16015.14
A16015.06EA 58.97 EUR
A16015.06 A16015.14
N-Acetonylpyridinium chloride ≥97%
N-Acetonylpyridinium chloride

Purity: 97% CAS No.: 42508-60-1 Molecular Formula: C8H10ClNO Formula Weight: 171.63 MDL No.: MFCD00031949

Vzorec: C₈H₁₀ClNO
Skladovacia teplota: Ambient
Číslo MDL: MFCD00031949
Číslo CAS: 42508-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR