Pentafluorophenyl isothiocyanate 96%
Dodávateľ: Thermo Fisher Scientific
|
Nebezpečí
|
L11874.03
L11874.06
L11874.03EA
88.65
EUR
L11874.03
L11874.06
Pentafluorophenyl isothiocyanate 96%
Pentafluorophenyl isothiocyanate
Purity: 96%
CAS No.: 35923-79-6
Molecular Formula: C7F5NS
Formula Weight: 225.14
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5378
Boiling Point: 109-110°/35mm
Beilstein Reference: 2736207
UN No.: 2922
MDL No.: MFCD00041042
Vzorec:
C₇F₅NS Teplota varu: 109…110 °C (35 mmHg) Hustota: 1,560 |
Číslo MDL:
MFCD00041042 Číslo CAS: 35923-79-6 EINECS: 252-791-9 ADR: 8,III |