Tlačiť

Methylethoxypyrazine (mixture of isomers), Sigma-Aldrich®

Dodávateľ: Merck


M-Clarity™ quality level = MQ300

W356905-100G-K W356905-1KG-K
SIALW356905-100G-KEA 364.22 EUR
SIALW356905-100G-K SIALW356905-1KG-K
Methylethoxypyrazine (mixture of isomers), Sigma-Aldrich®
Methylethoxypyrazine (mixture of isomers)
Vzorec: C₇H₁₀N₂O
Číslo CAS: 65504-94-1
EINECS: 265-794-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR