Tlačiť

Methyl-3-(methylthio)propionate, Sigma-Aldrich®

Dodávateľ: Merck

Synonymá: Methyl3-(methylthio)propionate

M-Clarity™ quality level = MQ300

W272000-1KG-K W272000-250G-K W272000-5KG-K
SIALW272000-1KG-KEA 279.58 EUR
SIALW272000-1KG-K SIALW272000-250G-K SIALW272000-5KG-K
Methyl-3-(methylthio)propionate, Sigma-Aldrich®
Methyl-3-(methylthio)propionate
Vzorec: C₅H₁₀O₂S
Číslo MDL: MFCD00008565
Číslo CAS: 13532-18-8
EINECS: 236-883-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR