Tlačiť

Thiocarbohydrazide, Sigma-Aldrich®

Dodávateľ: Merck

Synonymá: TCH

M-Clarity™ quality level = MQ100

223220-25G 223220-5G
SIAL223220-25GEA 231.12 EUR
SIAL223220-25G SIAL223220-5G
Thiocarbohydrazide, Sigma-Aldrich®
Thiocarbohydrazide
Vzorec: CH₆N₄S
Mw: 106.15 g/mol
Číslo MDL: MFCD00007616
Číslo CAS: 2231-57-4
EINECS: 218-769-8
OSN: 2811
ADR: 6.1 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR