Tlačiť

N,N-Dimethylcarbamoyl chloride 96%

Dodávateľ: Thermo Fisher Scientific
Nebezpečí

L03415.14 L03415.22
L03415.14EA 29.32 EUR
L03415.14 L03415.22
N,N-Dimethylcarbamoyl chloride 96%
N,N-Dimethylcarbamoyl chloride

Purity: 96% CAS No.: 79-44-7 Molecular Formula: C3H6ClNO Formula Weight: 107.54 Flash Point: 68°(154°F) Refractive Index: 1.4530 Odour: Unpleasant Melting Point: -33° Boiling Point: 167-168° Beilstein Reference: 878197 UN No.: 2262 MDL No.: MFCD00000635

Vzorec: C₃H₆ClNO
Teplota varu: 167…168 °C
Teplota tavenia: –33 °C
Hustota: 1,171
Teplota vzplanutia: 68 °C (154 °F)
Skladovacia teplota: Ambient
Číslo MDL: MFCD00000635
Číslo CAS: 79-44-7
EINECS: 201-208-6
OSN: 2262
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR