Tlačiť

N,N-Diisopropylcarbamoyl chloride, Sigma-Aldrich®

Dodávateľ: Merck


M-Clarity™ quality level = MQ100

365386-25G 365386-5G
SIAL365386-25GEA 222.62 EUR
SIAL365386-25G SIAL365386-5G
N,N-Diisopropylcarbamoyl chloride, Sigma-Aldrich®
N,N-Diisopropylcarbamoyl chloride
Vzorec: C₇H₁₄ClNO
Číslo MDL: MFCD00015013
Číslo CAS: 19009-39-3
EINECS: 242-743-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR