Tlačiť
Mini bottle-top dispensers, MINISPENSOR
Dodávateľ: witeg Labortechnik
5.371.525 5.371.990 5.371.950 5.371.101
613-9133EA 306.06 EUR
613-9133 WITG5.371.990 WITG5.371.950 WITG5.371.101
Mini bottle-top dispensers, MINISPENSOR
Dávkovače na lahve
This fix volume bottle dispenser is efficient for accurate delivery of microliter quantities.
  • For single handed operation
  • Suitable for safe storage in a fridge
  • Substitution for microliter pipettes
These dispenser are used for hospital laboratories for various tests like glucose, gama GT, creatinine, urea, cholesterol, amylase, lipose, iric acid, bilirubin.
Informace o dodávce: Supplied with a thread connector GL 28, 1 volume adapter and 1 bottle, both NS 29/32, suction tube and discharge tube.