Tlačiť
Automatic burettes, Schilling pattern
Dodávateľ: POULTEN GRAF
1.06832 1.06835 1.06839 1.06843 1.06845 1.06935 1.06939 1.06943 1.06945
POUL1.06832EA 97.87 EUR
POUL1.06832 POUL1.06835 POUL1.06839 POUL1.06843 POUL1.06845 POUL1.06935 POUL1.06939 POUL1.06943 POUL1.06945
Automatic burettes, Schilling pattern
Byrety
AR-GLAS®, clear or amber, class B.
  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe, reduced distance between subdivision marks
  • Automatic zeroing, squeezing bottle allows rapid filling, micrometre screw allows fine titration
Error limits according to class B, DIN EN ISO 385
Certifikácie: Error limits according to class B, DIN EN ISO 385