Tlačiť
Tubings, Original Perfusor®
Dodávateľ: BRAUN
8722935 8255172 8255059 8255253 8722919 8723060 8255067 8272565
720-2584EA 253.91 EUR
720-2584 BRAU8255172 BRAU8255059 BRAU8255253 BRAU8722919 BRAU8723060 BRAU8255067 BRAU8272565
Tubings, Original Perfusor®
Infuze
Original-Perfusor tubings for use with all syringe pumps inferior to four-bar pressure.
  • Minimized priming volumes
  • Luer-Lock fitting
  • Injection filter 0,22 μm and back-check valve for parallel infusions