Tlačiť

2'-Deoxyadenosine-5'-triphosphate disodium salt (dATP disodium salt) trihydrate, Sigma-Aldrich®

Dodávateľ: Merck

D6500-100MG D6500-10MG D6500-25MG
SIALD6500-100MGEA 602.89 EUR
SIALD6500-100MG SIALD6500-10MG SIALD6500-25MG
2'-Deoxyadenosine-5'-triphosphate disodium salt (dATP disodium salt) trihydrate, Sigma-Aldrich®
2'-Deoxyadenosine-5'-triphosphate disodium salt (dATP disodium salt) trihydrate
Vzorec: C₁₀H₁₄N₅Na₂O₁₂P₃·3H₂O
Skladovacia teplota: Freezer
Číslo CAS: 1927-31-7
EINECS: 277-809-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR