Tlačiť

2'-Deoxyguanosine monohydrate 99-100%, powder, BioReagent cell culture reagent, Sigma-Aldrich®

Dodávateľ: Merck

Synonymá: 2-Amino-9-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,9-dihydro-6H-purin-6-one hydrate

M-Clarity™ quality level = MQ200

D0901-100MG D0901-1G D0901-250MG
SIALD0901-100MGEA 152.83 EUR
SIALD0901-100MG SIALD0901-1G SIALD0901-250MG
2'-Deoxyguanosine monohydrate 99-100%, powder, BioReagent cell culture reagent, Sigma-Aldrich®
2'-Deoxyguanosine monohydrate
Vzorec: C₁₀H₁₃N₅O₄·H₂O
Skladovacia teplota: Ambient
Číslo MDL: MFCD00150760
Číslo CAS: 312693-72-4
EINECS: 213-505-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR