Tlačiť

Didodecyldimethylammonium bromide, Sigma-Aldrich®

Dodávateľ: Merck


M-Clarity™ quality level = MQ200

359025-10G 359025-2G
SIAL359025-10GEA 147.21 EUR
SIAL359025-10G SIAL359025-2G
Didodecyldimethylammonium bromide, Sigma-Aldrich®
Didodecyldimethylammonium bromide
Vzorec: C₂₆H₅₆BrN
Mw: 462.65 g/mol
Číslo MDL: MFCD00041969
Číslo CAS: 3282-73-3
EINECS: 221-923-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR