Tlačiť
Thermo trousers, Match 2023 MTH
Dodávateľ: FRISTADS KANSAS
1009225402XL 1009225403XL 1009225404XL 100922540L 100922540M 100922540S 100922540XL 100922540XS
KWIN1009225402XLEA 60.97 EUR
KWIN1009225402XL KWIN1009225403XL KWIN1009225404XL KWIN100922540L KWIN100922540M KWIN100922540S KWIN100922540XL KWIN100922540XS
Thermo trousers, Match 2023 MTH
Pracovní oděvy
Trousers made of 100% polyester. Suitable for use as underwear.
  • Adjustable waistband
  • Zip fly
Weight: 180 g/m²