Tlačiť

Terpinolene, Sigma-Aldrich®

Dodávateľ: Merck


M-Clarity™ quality level = MQ300

W304603-1KG-K W304603-9KG-K
SIALW304603-1KG-KEA 107.5 EUR
SIALW304603-1KG-K SIALW304603-9KG-K
Terpinolene, Sigma-Aldrich®
Terpinolene
Vzorec: C₁₀H₁₆
Číslo MDL: MFCD00049191
Číslo CAS: 586-62-9
EINECS: 209-578-0
OSN: 2541
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR