Tlačiť

Cacodylic acid (dimethylarsinic acid), Sigma-Aldrich®

Dodávateľ: Merck
Nebezpečí

Synonymá: Dimethylarsinic acid (Cacodylic acid)

M-Clarity™ quality level = MQ200

C0125-100G C0125-250G C0125-25G C0125-5G
SIALC0125-100GEA 978.97 EUR
SIALC0125-100G SIALC0125-250G SIALC0125-25G SIALC0125-5G
Cacodylic acid (dimethylarsinic acid), Sigma-Aldrich®
Cacodylic acid (dimethylarsinic acid)
Vzorec: (CH₃)₂As(O)OH
Mw: 138
Skladovacia teplota: Ambient
Číslo MDL: MFCD00002095
Číslo CAS: 75-60-5
EINECS: 200-883-4
OSN: 1572
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR