Cacodylic acid (dimethylarsinic acid) ≥98%
Dodávateľ: Thermo Fisher Scientific
|
Nebezpečí
|
Synonymá:
Dimethylarsinic acid (Cacodylic acid)
A12075.09
A12075.88
A12075.09EA
229.92
EUR
A12075.09
A12075.88
Cacodylic acid (dimethylarsinic acid) ≥98%
Cacodylic acid (dimethylarsinic acid)
Purity: 98%
CAS No.: 75-60-5
Molecular Formula: (CH3)2AsO2H
Formula Weight: 138.00
Melting Point: 200°
Beilstein Reference: 1736965
Merck Reference: 14,1604
UN No.: 1572
MDL No.: MFCD00002095
Vzorec:
(CH₃)₂As(O)OH Mw: 138 Skladovacia teplota: Ambient |
Číslo MDL:
MFCD00002095 Číslo CAS: 75-60-5 EINECS: 200-883-4 OSN: 1572 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...