Ammonium hexachlororhodate(III) hydrate ≥99.99% (metals basis), Premion®
Dodávateľ: Thermo Fisher Scientific
|
Nebezpečí
|
Synonymá:
Rhodium(III)-ammonium chloride hydrate
11812.03
11812.06
11812.03EA
981.77
EUR
11812.03
11812.06
Ammonium hexachlororhodate(III) hydrate ≥99.99% (metals basis), Premion®
Ammonium hexachlororhodate(III) hydrate
Purity: 99.99%
CAS No.: 15336-18-2
Molecular Formula: (NH4)3RhCl6.xH2O (x≈1) (formula not proven)
Formula Weight: 369.74(anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Dark red
Melting Point: dec.
MDL No.: MFCD00003394
Vzorec:
H₁₂Cl₆N₃Rh·H₂O Mw: 423.78 Teplota tavenia: Dec. Skladovacia teplota: Ambient |
Číslo MDL:
MFCD00003394 Číslo CAS: 15336-18-2 EINECS: 239-364-2 |