trans,trans-2,4-Undecadienal ≥90% remainder mainly trans,cis isomer
Dodávateľ: Thermo Fisher Scientific
Purity: 90+%
CAS No.: 30361-29-6
Molecular Formula: C11H18O
Formula Weight: 166.27
Storage Temperature: 2°C to 8°C
Flash Point: 107°(225°F)
Refractive Index: 1.5120
Boiling Point: 128-129°/10mm
Beilstein Reference: 1706330
MDL No.: MFCD00014677
Vzorec:
C₁₁H₁₈O Teplota varu: 128…129 °C (10 mmHg) Hustota: 0,870 Teplota vzplanutia: 107 °C (225 °F) |
Číslo MDL:
MFCD00014677 Číslo CAS: 30361-29-6 EINECS: 250-148-7 |