Tlačiť
Bottles, narrow neck, round, LDPE, with screw cap
Dodávateľ: LAMAPLAST
326G 326 324G 325 324 323G 323 319 317 325G
LAMA326GEA 11.34 EUR
LAMA326G 215-0166 LAMA324G 215-0149 215-0148 LAMA323G 215-0147 215-0146 215-0145 LAMA325G
Bottles, narrow neck, round, LDPE, with screw cap
Láhve Láhve plastové
LDPE, translucent or grey.
  • Leakproof, suitable for foodstuffs
  • Tagging points on shoulder and cap enables use of tamper evident tags and seals
  • Easy grip cap
  • Grey coloured for photosensitive products
Insert in cap ensures a good seal. Sloping shoulders reduce tendency for any residual liquids when pouring and emptying.
Pozor: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904