Tlačiť
Disposable earplugs, uvex com4-fit
Dodávateľ: UVEX
2112.004 2112.012
UVEX2112.004EA 68.19 EUR
UVEX2112.004 UVEX2112.012
Disposable earplugs, uvex com4-fit
Chrániče zátkové Chrániče zátkové jednorázové
Thanks to their compact design, the com4-fit earplugs are ideal for wearers with narrow ear canals.
  • Ergonomic shape adapts to the natural shape of the ear canal
  • Combines optimal comfort with the highest level of protection
  • x-grip technology reduces expansion pressure and therefore increases wearer comfort, even when worn for extended periods
  • Easy insertion and removal
  • For small to medium ear canals
  • High visibility colour for easy visual recognition
SNR: 33 dB
Certifikácie: EN 352-2:2002