Tlačiť
RT-PCR kits, Verso™
Dodávateľ: Thermo Fisher Scientific
AB-1453/A AB-1453/B AB-1454/LD/B
733-1097EA 452.47 EUR
733-1097 733-1098 733-1103
RT-PCR kits, Verso™
Činidla pro nukleové kyseliny qPCR/RT-qPCR Enzymes and Kits
The Verso™ RT-PCR system combines a reverse transcription enzyme, improved priming options and optimised buffers to generate high yield and full length cDNA. With a variety of kit options for both 1- and 2-step RT-PCR reactions, Verso™ provides optimum flexibility and sensitivity in an easy to use format.