Tlačiť
GelRed® & GelGreen® Nucleic acid gel stains
Dodávateľ: Biotium
41001 41002 41003 41004 41003-1 41005 41005-1 41003-T 41002-1
41001.EA 451.43 EUR
41001. 41002. 41003. 41004. 41003-1 730-1535 41005-1 41003-T 41002-1
GelRed® & GelGreen® Nucleic acid gel stains
Barviva Barviva pro elektroforézu
GelRed and GelGreen are highly sensitive DNA/RNA gel stains made safe by preventing penetration into cells.
  • Highly sensitive either as precast gel stains or post gel stains
  • Can be microwaved and stored at room temperature
  • GelRed™ has been shown to be less mutagenic than ethidium bromide
  • No destaining necessary
GelRed is compatible with any standard 312 nm transilluminator.
GelGreen can be used with a 254 nm UV transilluminator or gel reader with visible light excitation.