Tlačiť
Media supplements for dehydrated culture media
Dodávateľ: Merck
1.04039.0001 1.00092.0010 1.00093.0010 1.09877.0010 1.09875.0010 1.00898.0010 1.00051.0010 1.00079.0010 1.12122.0010 1.11781.0010 1.07627.0010 1.07624.0010 1.07006.0010 1.09874.0010 1.00045.0010 1.00888.0010 1.00071.0010 1.08981.0100
1.04039.0001EA 63.78 EUR
1.04039.0001 1.00092.0010 1.00093.0010 1.09877.0010 1.09875.0010 1.00898.0010 1.00051.0010 1.00079.0010 1.12122.0010 1.11781.0010 1.07627.0010 1.07624.0010 1.07006.0010 1.09874.0010 1.00045.0010 1.00888.0010 1.00071.0010 1.08981.0100
Media supplements for dehydrated culture media
Média mikrobiologická Mikrobiologické přísady a doplňky
The isolation of microorganisms often requires the use of selective media. The use of freeze-dried supplements ensures an optimal and standardised medium preparation and thus optimised performance of the selective medium.
  • No aerosols released upon opening, therefore minimising inhalation of toxic agents
  • User friendly screw caps which guarantee easy and safe opening
  • Standard volumes make preparation easier