Tlačiť
Headspace vials, WHEATON®
Dodávateľ: DWK LIFE SCIENCES
225277 225280 W225282
WHEA225277EA 67.39 EUR
WHEA225277 WHEA225280 WHEAW225282
Headspace vials, WHEATON®
Vialky
These vials made of type I borosilicate glass feature rounded shoulders and bottoms, allowing for even heating and safer operation at higher pressures.
  • Accepts 20 mm aluminium seal
  • Top designed to provide a tight seal with septa
Objednávacie informácie: Aluminium seals must be ordered separately.