Tlačiť
Micro surface stirrer shafts, BOLA
Dodávateľ: Bohlender
C486-20 C486-16 C486-12 C486-08
BOHLC486-20EA 143.2 EUR
BOHLC486-20 BOHLC486-16 BOHLC486-12 BOHLC486-08
Micro surface stirrer shafts, BOLA
Míchadla Hřídele míchací
Micro stirrer shafts made of PTFE-jacketed stainless steel with blade made completely of PTFE. With four round paddles.
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C
Ideal for stirring in test tubes or narrow-mouth vessels, especially suitable for mixing in round vessels and in vessels with low fill level.