Tlačiť
UV/white light transilluminators
Dodávateľ: UVP ULTRA VIOLET PRODUCTS
95-0415-02 95-0415-05 95-0418-02
732-4370EA 2570 EUR
732-4370 732-4371 732-4373
UV/white light transilluminators
Transiluminátory Transiluminátory s UV/bílým světlem
These UV/white light transilluminators house white light and UV working areas side-by-side. White light can be used for viewing Coomassie Blue stained gels, silver stained protein gels, autoradiographs and microtitre plates. The UV side can be used for viewing ethidium bromide gels.
  • Easily switch between light sources using the rocker switch on the front of the unit
  • UV side utilises UVG™ long-life filter
  • Clear UV blocking cover is hinged and can be height adjusted to provide a suitable viewing angle or to access the filter surface
W×D×H: 486×337×143 mm (height including UV blocking cover)
Weight: 9,6 kg