Tlačiť
Precision PT100 reference sensors
Dodávateľ: JULABO GMBH
8902213 8902215 8902223 8902225 8981002
461-3533EA 799.52 EUR
461-3533 461-3534 461-3535 461-3536 461-3537
Precision PT100 reference sensors
Elektrody Elektrody ostatní
Sensor is suitable for use with calibration baths.
  • Integrated measuring electronics and RS232 connection
  • Measurement range −30 to +300 °C
  • Measurement accuracy is ±0,05 °C over measurement range, except from +0,01 to 100 °C where accuracy is ±0,01 °C
Dimensions: 180×4 mm