Tlačiť

Phosphorus(V) fluoride

Dodávateľ: abcr

AB104073.0250 AB104073.0050
ABCRAB104073.0250EA 0 EUR
ABCRAB104073.0250 ABCRAB104073.0050
Phosphorus(V) fluoride
Phosphorus(V) fluoride
Vzorec: F₅P
Teplota tavenia: –83 °C
Hustota: 5,805 g/cm³ (20 °C)
Číslo MDL: MFCD00040326
Číslo CAS: 7647-19-0
EINECS: 231-602-3
OSN: 2198
ADR: 2,
Merck Index: 12,07509

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR