Tlačiť
Cleanroom gloves, latex, sterile, BioClean Advance™
Dodávateľ: NITRITEX
BASL6.0 BASL7.0 BASL8.0 BASL9.0 BASL7.5 BASL8.5 BASL6.5
112-3492EA 546.33 EUR
112-3492 112-3494 112-3496 112-3498 112-3495 112-3497 112-3493
Cleanroom gloves, latex, sterile, BioClean Advance™
Rukavice
These hand-specific latex gloves are anatomically shaped and therefore comfortable to wear. They can be used in medical areas as well as in laboratories.
  • Sterile, powder-free
  • Latex provides exceptional flexibility and comfort
  • Protein level: less than 50 µg/g
  • Double-donnable
  • Textured surface for good grip
  • Anatomically shaped for enhanced wearer comfort
  • Cuff marking: Ink Print Size and L or R
  • AQL 1,5
  • Standard compliance: PPE, Cat III EN ISO 374 Type B
Suitable for use in Class 10, ISO 4 or M2.5 cleanrooms.
EN 455, EN 374, Cat. III; AQL 1.5
Certifikácie: Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 +A1:2009 and EN 374-1:2016, EN 374-5:2016 and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Sterilized in accordance with ISO 11137-1:2006.
Suitable for contact with dry, alcoholic and acidic foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Balenie: One cuffed pair per plastic wallet; one wallet per EasyTear™ PE pouch; 10 pairs per sealed outer PE bag; 20 outer bags per carton liner; one carton liner per carton (200 pairs).