Tlačiť
Hose tubing connectors, straight, cylindrical nozzles, LaboPlast®
Dodávateľ: BURKLE
8701-0008 8701-0004 8701-0006 8701-0010 8701-0012 8701-0014 8710-0012 8710-0004 8710-0006 8710-0008 8710-0010 8710-0014
229-7434EA 6.9 EUR
229-7434 229-0245 229-0246 229-0247 229-0248 229-0249 229-0223 229-0218 229-0219 229-0220 229-0221 229-0224
Hose tubing connectors, straight, cylindrical nozzles, LaboPlast®
Fitinky a konektory
Cylindrical straight hose connectors for various hose sizes. PP or PVDF, translucent.
  • PP, temperature resistant up to +135 °C max.
  • PVDF, temperature resistant up to +160 °C max., highly resistant to chemicals
  • Sterilisable
PVDF connectors with food industry approval, autoclavable and UV-resistant.