Tlačiť
Wire baskets, rectangular, with lid
Dodávateľ: HUPFER METALLWERKE
7501132 7501135 7501133 7501204 7501161 7501162 7501282
HUPF7501132EA 291.62 EUR
HUPF7501132 HUPF7501135 HUPF7501133 HUPF7501204 HUPF7501161 HUPF7501162 HUPF7501282
Wire baskets, rectangular, with lid
Koše a košíky Koše drátěné
These fine mesh baskets, made of stainless steel, electropolished, are designed to keep small parts and accessories.
  • With lid
  • With base frame
  • Mesh size: 1,6×1,6 mm
Colour: Silver.