Tlačiť
Measuring beakers with handle
Dodávateľ: BURKLE
7104-3000 7104-5000 7104-0500 7104-1000 7104-2000 7104-0250 7102-0100 7102-0500 7102-1000 7102-2000 7102-5000 7102-0250
BURK7104-3000EA 22.26 EUR
BURK7104-3000 BURK7104-5000 BURK7104-0500 BURK7104-1000 BURK7104-2000 BURK7104-0250 BURK7102-0100 BURK7102-0500 BURK7102-1000 BURK7102-2000 BURK7102-5000 BURK7102-0250
Measuring beakers with handle
Kádinky
PP, highly transparent.
  • Ergonomic handle for secure grip
  • Functional spout to minimize spills
  • Cleaning up to max. +60 °C is recommended to preserve marks and inscriptions, for autoclaving (+121 °C) beakers with an transparent scale should be selected
Ideal for general laboratory use and a wide variety of other applications.