Tlačiť
Microcentrifuge tubes, 0,5/1,5/1,8 ml, Micrewtube®
Dodávateľ: SIMPORT SCIENTIFIC
T341-4T T341-6T T341-7T T341-2T T341-5T T341-4TBR T341-2TLST T341-6TPR T341-6TBR T341-4TPR T341-2TBR T341-2TPR T341-4TLST T341-5TBR T341-5TLST T341-5TPR T341-6TLST T341-7TBR T341-7TLST T341-7TPR
612-6651EA 103.49 EUR
612-6651 612-6655 612-6650 612-6653 612-6654 SIMPT341-4TBR SIMPT341-2TLST SIMPT341-6TPR SIMPT341-6TBR SIMPT341-4TPR SIMPT341-2TBR SIMPT341-2TPR SIMPT341-4TLST SIMPT341-5TBR SIMPT341-5TLST SIMPT341-5TPR SIMPT341-6TLST SIMPT341-7TBR SIMPT341-7TLST SIMPT341-7TPR
Microcentrifuge tubes, 0,5/1,5/1,8 ml, Micrewtube®
Zkumavky Mikrozkumavky centrifugační
PP, translucent or PP dark brown.
  • Available in plain or graduated configuration
  • Max. RCF: Non skirted tubes can be centrifuged up to 20 000×g
  • Temperature range: –196 to +121 °C
  • Certified RNase-, DNase-, pyrogen- and DNA-free