Tlačiť
GL bottle septa, silicone rubber, DURAN®
Dodávateľ: DWK LIFE SCIENCES
292460503 292460606 292460803 292460906 292461002
SCOT292460503EA 113.12 EUR
SCOT292460503 SCOT292460606 SCOT292460803 SCOT292460906 SCOT292461002
GL bottle septa, silicone rubber, DURAN®
Septum
Silicone, translucent.
  • Ideal for use as septa (for piercing)
  • Suitable for PBT screw cap with aperture
  • Good thermal resistance (130 °C vapour/200 °C dry heat)
For threaded connection caps with GL thread.