Tlačiť
HPLC kolony, Accucore™ PFP
Dodávateľ: Thermo Fisher Scientific
17426-012105 17426-013005 17426-014005 17426-052130 17426-053030 17426-102130 17426-103030 17426-104630 17426-152130 17426-153030 17426-154630
554-1954EA 640.19 EUR
554-1954 554-1955 554-1956 554-1921 554-1922 554-1924 554-1925 554-1926 554-1927 554-1928 554-1929
HPLC kolony, Accucore™ PFP
Kolony chromatografické
These columns offer alternative selectivity to C18, particularly for halogenated analytes. Based on Core Enhanced Technology, they provide fast, high resolution separations without the elevated back pressures required by sub-2 µm particles.
  • Lower back pressures
  • Extra retention for halogenated species
  • Unique selectivity for non halogenated polar compounds