Tlačiť
Test tube racks, rectangular
Dodávateľ: BOCHEM
8210 8212 8214 8220 8222
212-0001EA 49.74 EUR
212-0001 212-0002 212-0003 212-0004 212-0005
Test tube racks, rectangular
Stojany Tube Racks
Rack, made of stainless steel 18/10.
  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean