Tlačiť
Tissue cassettes in QuickLoad™ sleeves, Swingsette™, M517SL
Dodávateľ: SIMPORT SCIENTIFIC
M517-10SL M517-11SL M517-12SL M517-2SL M517-3SL M517-4SL M517-5SL M517-6SL M517-7SL M517-8SL M517-9SL
720-1471EA 204.97 EUR
720-1471 720-1472 720-1473 720-1477 720-1478 720-1479 720-1480 720-1481 720-1482 720-1483 720-1484
Tissue cassettes in QuickLoad™ sleeves, Swingsette™, M517SL
Kazety pro zalévání vzorků Kazety tkáňové
Acetal polymer.
  • Hinged lid supplied in separate packaging to make it easier to use the cassettes with commercial printers; large tab to make opening easier
  • Disposable plastic cassettes that are highly resistant to histological solvents
  • Oblong openings for good fluid exchange
  • Front face inclined 45° so as to be usable with certain automatic labelling systems
Balenie: Each case contains 10 sleeves of 75 cassettes and 10 bags of 75 lids.