Tlačiť
Sample dippers
Dodávateľ: BURKLE
5315-0050 5315-0200 5315-0100 5315-0300
BURK5315-0050EA 66.59 EUR
BURK5315-0050 331-2113 BURK5315-0100 BURK5315-0300
Sample dippers
Vzorkovače Naběračky na vzorky
Plunging siphons for the simple removal of liquid samples.
  • Made from transparent PP
  • All-layer and point sampling
  • Upper end with reflux valve, lower end with ball valve