Tlačiť
Stirrer blades for balloon flasks
Dodávateľ: KARTELL
690 691 692 693 694
KART690EA 28.6 EUR
KART690 KART691 KART692 KART693 KART694
Stirrer blades for balloon flasks
Míchadla Čepele míchací
Fit all standard button tip stirring shafts. 'Long groove' keyhole (Ø 9 mm) provides easy insertion into flask neck and proper position for stirring action. Can be used continuously at temperatures up to 250 °C and, for short intervals, to 300 °C.